Learn More
Specifications
Specifications
| Physical Form | Crystalline |
| Quantity | 12 kg |
| Linear Formula | (NH4)2SO4 |
| Formula Weight | 132.14 |
| Percent Purity | ≥99.0% |
| Grade | ACS |
| Packaging | Poly Pail |
| Density | 1.77g/cm3 (20°C); Bulk Density: 850 kg/m3 |
| Chemical Name or Material | Ammonium Sulfate, 99.0%, Suprapur™ |
Safety and Handling
| Recommended Storage | Store without restrictions |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.