Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Sulfate, MACCO

Catalog No. 502985631
Encompass
Change view
Click to view available options
Quantity:
12 kg
50 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-298-5631 12 kg
50-298-5653 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-298-5631 Supplier Thermo Scientific Production Chemicals and Services Supplier No. M50704000120
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7783-20-2
Molecular Weight (g/mol) 132.14 g/mol

Specifications

Physical Form Fine Crystals or Crystalline Powder
Quantity 12 kg
Percent Purity 100%
Grade USP
Packaging 12 kg poly drum Curtec + 2 x PE liner inside
Chemical Name or Material Ammonium Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.