Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Ammonium Sulfate, Molecular Biology Grade, Calbiochem™,

Catalog No. 1683561KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-835-61KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-835-61KG Supplier MilliporeSigma™ Supplier No. 1683561KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity reagent suitable for various molecular biology applications, including protein precipitation.

Hygroscopic
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Physical Form Crystalline
Quantity 1 kg
Assay Percent Range ≥99%
Solubility Information Soluble in water
Formula Weight 132.1
Percent Purity ≥99%
Grade by acidometry
Chemical Name or Material Ammonium Sulfate
RTECSNumber BS4500000
Recommended Storage Store at 15° to 30°C (59° to 86°F).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.