Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Sulfate, MP Biomedicals™

Catalog No. MP219140905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219140905 5 kg
MP021914095 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219140905 Supplier MP Biomedicals, Inc Supplier No. 0219140905
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for the precipitation or fractionation of proteins and purification of antibodies. Ammonium Sulfate, MP Biomedicals is an inorganic salt useful in crystallographic analysis of nucleic acids and proteins. Use in long PCR buffer, PCR lysis solution and

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Physical Form Powder
Quantity 5 kg
Solubility Information Soluble freely in water; insoluble in alcohol, acetone.
Formula Weight 132.1
Percent Purity 99%
Grade ACS
Chemical Name or Material Ammonium Sulfate
Recommended Storage Room Temperature, (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.