Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Sulfate, MP Biomedicals™

Catalog No. MP04808229 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04808229 5 kg
MP04808210 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP04808229 Supplier MP Biomedicals, Inc Supplier No. 04808229
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium Sulfate, ltra Pure Grade, ≥99%, White Crystalline Powder

  • Ammonium sulfate is a widely used reagent in molecular biology and chromatography.
  • It is useful in the isolation and purification of enzymes and proteins.
  • It is used for the precipitation or fractionation of proteins or for purification of antibodies.
  • It is also useful for crystallographic analysis of nucleic acids and proteins
  • Ammonium sulfate is also widely used in HPLC of proteins, such as in hydrophobic interaction chromatography
  • Ammonium sulfate is used in long PCR buffer, in PCR lysis solution, and in secondstrand cDNA synthesis.
  • Also it is used for the HPLC analysis of phospholipids.
  • Ammonium sulfate is an inorganic salt prepared by treating ammonia with sulfuric acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym Sulfuric acid diammonium salt, Mascagnite
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Melting Point >280°C (decomposition)(literature)
Color White
pH 5 to 6 (5% aq soln)
Physical Form Crystalline Powder
Quantity 5 kg
Solubility Information Soluble freely in water; insoluble in alcohol, acetone.
Percent Purity ≥99%
Grade Ultra Puregrade
Residue after Ignition 8e-006
Density 1.77g/mL at 25°C (Lit.)
Chemical Name or Material Ammonium Sulfate
Show More Show Less
RTECSNumber BS4500000
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.