Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Ammonium Sulfate OmniPur™ Calbiochem™,

Catalog No. 21605KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-605-KG 5 kg
21-702-5KG 25 kg
21-501-KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-605-KG Supplier MilliporeSigma™ Supplier No. 21605KG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Quantity 5 kg
Assay Percent Range ≥99.5%
Solubility Information Soluble in water
Formula Weight 132.1
Chemical Name or Material Ammonium Sulfate
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.