Learn More
Specifications
Specifications
| Melting Point | 149°C |
| Color | White |
| pH | 5 to 6 |
| Physical Form | Solid |
| Quantity | 3 kg |
| Assay Percent Range | ≥97.5 % |
| Linear Formula | NH4SCN |
| Formula Weight | 76.12 |
| Percent Purity | ≥97.5% |
| Grade | Certified ACS |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Contact with acids liberates toxic gas. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. NFPA Health:2 Flammability:1 Instability:1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.