Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Thiocyanate, Purified, Reagents

Catalog No. 01337483
Change view
Click to view available options
Quantity: 500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-337-483 Quantity: 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-483 Supplier Reagents Holdings Llc Supplier No. C2139500500A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium Thiocyanate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.12
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym Ammonium isothiocyanate
IUPAC Name ammonium cyanosulfanide
SMILES [NH4+].[S-]C#N

Specifications

Boiling Point 170°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Poly Bottle
Specific Gravity 1.3
Chemical Name or Material Ammonium Thiocyanate, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.