Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium thiocyanate solution, Volumetric, 0.1 M NH4SCN (0.1N), Solstice

Catalog No. 6002588 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-025-88 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-025-88 Supplier Solstice Supplier No. 350361L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Volumetric, 0.1 M NH4SCN (0.1N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1762-95-4 , 7732-18-5
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
MDL Number MFCD00011428
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Boiling Point 100°C
CAS Min % ≥99.0000%
CAS Max % ≤1.0000%
Quantity 1 L
Linear Formula NH4SCN
Beilstein 3595135
Formula Weight 76.12
Concentration or Composition (by Analyte or Components) 0.1 M NH4SCN (0.1N)
Packaging Glass Bottle
Chemical Name or Material Ammonium thiocyanate solution, Volumetric, 0.1 M NH4SCN (0.1N)
ShelfLife 720 days from date of manufacture
EINECSNumber 231-791-2
Recommended Storage Room Temp
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.