Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium trifluoromethanesulfonate, 99%

Catalog No. AC388730250
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC388730250 25 g Glass bottle
AC388730050 5 g Glass bottle
AC388730500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC388730250 Supplier Thermo Scientific Chemicals Supplier No. 388730250
Only null left
Add to Cart

CAS: 38542-94-8 | CH4F3NO3S | 167.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38542-94-8
Molecular Formula CH4F3NO3S
Molecular Weight (g/mol) 167.11
MDL Number MFCD00075332
InChI Key BMWDUGHMODRTLU-UHFFFAOYSA-N
Synonym ammonium trifluoromethanesulfonate, ammonium triflate, acmc-20alsc, methanesulfonic acid,1,1,1-trifluoro-, ammonium salt 1:1
PubChem CID 15842047
IUPAC Name azanium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].[NH4+]

Specifications

Melting Point 225°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99%
Formula Weight 167.11
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Ammonium trifluoromethanesulfonate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.