Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium zinc sulfate hydrate

Catalog No. AAA135840B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA135840B 1000 g
AAA1358430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA135840B Supplier Thermo Scientific Chemicals Supplier No. A135840B
Add to Cart

CAS: 77998-33-5 | H8N2O8S2Zn | 293.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77998-33-5
Molecular Formula H8N2O8S2Zn
Molecular Weight (g/mol) 293.57
MDL Number MFCD18711942
InChI Key CHORECZHRAUKSW-UHFFFAOYSA-L
Synonym ammonium zinc sulfate hydrate, acmc-20alsa, ammonium hydrate zinc sulfate, cvgyqralnwebmp-uhfffaoysa-m, ammonium ? 1-zinc 1+ hydrate sulfate, sulfuric acid, ammoniumzinc salt 2:2:1 , hydrate 9ci
PubChem CID 53471899
SMILES [NH4+].[NH4+].[Zn++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Quantity 1000 g
UN Number UN3077
IUPAC Name azanium;zinc;sulfate;hydrate
Formula Weight 293.57 (Anhydrous)
Density 1.931
Chemical Name or Material Ammonium zinc sulfate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.