Learn More
Abcam AMN082 Dihydrochloride, mGlu7 Agonist, Solid, 10 mg
Description
- Molecular Weight: 465.5 Da
- Purity: >98%
- Form: Solid
- Activity: Potent, selective, orally active and brain penetrant mGlu7 receptor agonist
- Experimental Data: Potently inhibits cAMP accumulation and stimulates GTP binding (EC50 = 64–290 nM) in mGluR7 transfected cells; selective over mGlu1, 2, 3, 4, 5, 6, and 8, and ionotropic receptors (up to 10 μM)
- In Vivo Effects: Elevates plasma stress hormones; induces mGlu7 receptor internalisation in dissociated hippocampal neurones
- Storage Conditions: Store at +4°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | mGlu7 Receptor |
| Form | Solid |
| Shipping Condition | Ambient |
| Buffer | 100% AMN082 |
| Content And Storage | Store at 4°C. |
| Quantity | 10 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.