Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam AMN082 Dihydrochloride, mGlu7 Receptor Agonist, Solid, 50 mg

Catalog No. 502707943
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7943 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7943 Supplier Abcam Supplier No. AB12001150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent, selective, orally active and brain penetrant mGlu7 receptor agonist.

  • Molecular Weight: 465.5 Da
  • Purity: >98%
  • Form: Solid
  • Activity: Potent, selective, orally active and brain penetrant mGlu7 receptor agonist
  • Mechanism: Potently inhibits cAMP accumulation and stimulates GTP binding (EC50 = 64–290 nM) in mGluR7 transfected cells
  • Selectivity: Selective over mGlu1, 2, 3, 4, 5, 6, and 8, and ionotropic receptors (up to 10 μM)
  • Biological effects: Elevates plasma stress hormones in vivo; induces mGlu7 receptor internalisation in dissociated hippocampal neurones
  • Citation Count: 11

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule mGlu7 Receptor
Content And Storage Store at 4°C.
Form Solid
Shipping Condition Ambient
Buffer 100% AMN082
Formulation 100% AMN082
Quantity 50 mg
For Use With (Application) mGlu7 receptor research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.