Learn More
Invitrogen™ Amnionless Monoclonal Antibody (260808)

Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | Amnionless |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 260808 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Amn |
| Gene Accession No. | Q9BXJ7 |
| Gene Alias | 5033428N14Rik; Amn; amnion associated transmembrane protein; amnionless; amnionless homolog; AV002116; PRO1028; Protein amnionless; Soluble protein amnionless; type I transmembrane protein; UNQ513/PRO1028; visceral endoderm-specific type 1 transmembrane protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.