Learn More
Sigma Organic Chemistry Ammonium zirconium(IV) carbonate solution | 250ML | MFCD00046155
Supplier: Sigma Organic Chemistry 464597250ML
Ammonium zirconium(IV) carbonate solution, 250ML
About This Item:
MDL Number: MFCD00046155
UNSPSC Code: 12352302
Storage: 2-8C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.