Learn More
Bioworld Amoxicillin + Potassium Clavulanate, 2 g
Supplier:  Bioworld 401100121
Encompass_Preferred
Amoxicillin, Potassium Clavulanate Inhibitor of bacterial cell wall synthesis with high activity against Gram-negative bacteria. Specifications:Mixture contains 5 to 1 amoxicillin to potassium clavulanate.Cell culture tested.β-lactamase sensitive.May cause sensitization by inhalation and skin contact.For in vitro lab use only. Not for human use or consumption.
Catalog No. 50-255-299
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.