Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amoxicillin Trihydrate 98.0+%, TCI America™
SDP

Catalog No. A209925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A209925G 25 g
A20995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A209925G Supplier TCI America Supplier No. A209925G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61336-70-7
Molecular Formula C16H25N3O8S
Molecular Weight (g/mol) 419.449
MDL Number MFCD00072029
InChI Key MQXQVCLAUDMCEF-FXHJIJKUSA-N
PubChem CID 124080955
IUPAC Name (2S,5R,6S)-6-[[(2S)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;trihydrate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)O)C.O.O.O

Specifications

Physical Form Powder/Crystals
Color White/Yellow
Quantity 25 g
Formula Weight 365.40
Percent Purity ≥98.0% (T)
Chemical Name or Material Amoxicillin Trihydrate
RTECSNumber XH8300000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.