Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AMPA ReceptorTocris Bioscience™et™

Catalog No. 18221R
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-221-R 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-221-R Supplier Tocris Bioscience™ Supplier No. 1822/1 SET
This item is not returnable. View return policy

Selection of 5 AMPA receptor ligands (Cat. Nos. 0254, 0306, 1044, 0713 and 1454)

Contains the listed products as lyophilised solids which can be used to investigate the pharmacology of AMPA receptors.

Specifications

Content And Storage -20°C
Quantity 1 Set
Product Type AMPA Receptor Tocriset™
Inhibitors AMPA Receptor Tocriset™

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.