Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Ampere's Rule Apparatus
SDPSCI_ED

Catalog No. S139479 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139479 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139479 Supplier United Scientific™ Supplier No. AMPR01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Classic and easy to use demonstration.

This apparatus allows students to study the magnetic field around a wire.

  • A heavy brass wire with terminals is arranged on a clear plastic base
  • Also included are one each 45 mm compass, six each 16 mm compasses, and an activity guide

Specifications

Grade 5 to 12
Format Kit
Languages English
Subject Physics

Warning: Cancer and reproductive harm

Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.