Learn More
TARGETMOL CHEMICALS INC AMPEROZIDE 5MG
Supplier: TARGETMOL CHEMICALS INC T23733
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Amperozide (FG 5606) is a diphenylbutylpiperazine atypical antipsychotic which antagonizes 5-HT2A receptors. It inhibits dopamine release and alters the firing of dopaminergic neurons. Investigations regarding the use of the agent revolved primarily around its capability for treating schizophrenia in humans. purity: 99%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.