Learn More
AmpFLSTR™ PCR Enhancer
Description
AmpFLSTR™ PCR Enhancer is a reagent that enhances PCR amplification when used with buccal samples collected using the Bode Buccal DNA Collector™ and processed using the GlobalFiler™ Express PCR Amplification Kit. By optimizing primer binding efficiency, PCR Enhancer helps simplify data analysis and reduce time to result.
- Optimized for Bode Buccal DNA Collector samples treated with Prep-n-Go™ Buffer and processed using the GlobalFiler Express PCR Amplification Kit
- Simply add PCR Enhancer to the PCR reaction reagents during amplification setup without pretreatment or incubation steps
- Helps improve first pass success when using the GeneMapper™ ID-X automated genotyping expert software system for data analysis
AmpFLSTR PCR Enhancer incorporates easily into the existing direct amplification workflows of forensic laboratories processing single-source samples.
Specifications
Specifications
| Quantity | 500 reactions |
| Product Line | AmpFLSTR |
| Polymerase | Taq DNA Polymerase |
| Product Type | PCR Enhancer |
| Shipping Condition | Approved for shipment on dry ice |
| Content And Storage | 1.25 mL AmpFLSTR PCR Enhancer Store at -5 to -30°C on receipt, 2 to 8°C after initial use. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.