Learn More
Cerilliant Corporation AMPHETAMINE D-11 1ML
Supplier:  Cerilliant Corporation A019
Encompass_Preferred
Amphetamine D-11 | 66432-30-2 | MW 146.27 | 1ml | Amphetamine, a stimulant of the phenethylamine class, is found in ADHD-treatment drugs such as Adderall, Vyvanse, and ProCentra. Because of its stimulant effect, amphetamine is used recreationally as a performance-enhancer and illicit drug. This stable-labeled internal standard is suitable for quantitation of amphetamine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or sports testing.
Catalog No. NC2326128
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.