Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Amphiregulin Monoclonal Antibody (AREG559), APC, eBioscience™, Invitrogen™

Catalog No. p-7138216
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-537-042 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-537-042 Supplier Invitrogen™ Supplier No. 17537042
Only null left
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: The AREG858 monoclonal antibody reacts with human Amphiregulin (AR). AR is a member of the Epidermal Growth Factor (EGF) family and is expressed as a transmembrane precursor protein and released by proteolytic cleavage. Soluble AR is a ligand for the EGF receptor (EGFR) and the interaction promotes growth of normal epithelial cells, fibroblasts and keratinocytes. In the context of the immune system AR functions as an effector molecule for tissue repair and homeostasis, mediates resistance and tolerance to infection, and suppresses inflammation. Under different disease and activating conditions AR is expressed in T cells, group 2 innate lymphoid cells, basophils, mast cells, eosinophils, and neutrophils. Applications Reported: This AREG559 antibody has been reported for use in flow cytometric analysis. Applications Tested: This AREG559 antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (Product No. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

Amphiregulin (AR) binds to EGF-Receptor (EGFR) with lower affinity than EGF. The mature secreted form AR is an 84-amino acid residue glycosylated polypeptide growth regulator, which is generated by proteolytic processing of a 252-amino acid transmembrane precursor. Seven different polypeptide ligands, which derive from distinct genes, are capable of binding to the extracellular domain of EGFR. These ligands include EGF, TGFalpha, AR, HB-EGF, cripto-1, epiregulin, and betacellulin. All of these growth factors contain a characteristic EGF-like domain which is defined by 6 evenly spaced cysteine residues that generate 3 loops through the formation of disulfide bonds. AR protein has been localized by immunohistochemistry to the epithelium of the colon, stomach, pancreas, breast, and placenta. AR is reportedly overexpressed in human cancers of breast, colon, stomach, and pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amphiregulin
Applications Flow Cytometry
Classification Monoclonal
Clone AREG559
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene AREG
Gene Accession No. P15514
Gene Alias amphiregulin; amphiregulin B; AR; AREG; AREGB; colorectum cell-derived growth factor; CRDGF; Mcub; MGC13647; schwannoma-derived growth factor; SDGF
Gene Symbols AREG
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.