Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampicillin, Anhydrous, 98.24%, MP Biomedicals™

Catalog No. ICN19014605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19014605 5 g
ICN19014625 25 g
ICN19014680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19014605 Supplier MP Biomedicals, Inc Supplier No. 0219014605

Chemical Identifiers

CAS 69-53-4
Molecular Formula C16H19N3O4S
Molecular Weight (g/mol) 349.405
InChI Key AVKUERGKIZMTKX-NJBDSQKTSA-N
Synonym Aminobenzylpenicillin
PubChem CID 6249
ChEBI CHEBI:28971
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C

Specifications

Type Ampicillin, Anhydrous
Physical Form Powder
Color White
Quantity 5 g
Assay Percent Range 0.9824
Formula Weight 371.4
Chemical Name or Material Ampicillin, Anhydrous, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.