Learn More
Cayman Chemical Ampicillin (sodium salt), 69-52-3, 100 g
SDP
Supplier:  Cayman Chemical 14417100
Encompass_Preferred
Molecular Formula C16H18N3O4S • Na, Purity ≥95%, Formula Weight 371.4, Synonyms: (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, monosodium salt; A broad-spectrum antibiotic; active against Gram-positive and Gram-negative bacteria, including veterinary isolates of S. pseudintermedius, S. aureus, E. coli, Pasteurella, and S. canis (MIC50s = 0.25, 0.5, 2, 0.12, and 0.25 g/ml, respectively); reduces cough frequency, tachypnea, dyspnea, and fever and increases survival in a baboon (P. cynocephalus) model of pneumococcal pneumonia at 80 mg/kg.
Catalog No. NC0979673
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.