Learn More
Sigma Aldrich Fine Chemicals Biosciences Ampicillin, 10 mL
Supplier: Sigma Aldrich Fine Chemicals Biosciences A535410ML
Ready Made Solution, 100 mg/mL, 0.2 μm filtered. Ampicillin is used to select bacteria cells with specific resistance during general microbiology studies. It is used to study antibiotic resistance and penetration limitations, the synergy between multiple antibiotics, certain bloodstream infections, and has been used to develop PCR assays to detect resistance genes in cerebrospinal fluid.