Learn More
Invitrogen™ AMPK gamma-2 Polyclonal Antibody

Description
Recommended positive controls: HepG2, Mouse brain. Predicted reactivity: Mouse (98%), Rat (97%), Chicken (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | AMPK gamma-2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.96 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | PRKAG2 |
| Gene Accession No. | Q91WG5, Q9UGJ0 |
| Gene Alias | 2410051C13Rik; 5'-AMP-activated protein kinase subunit gamma-2; 5-AMP-activated protein kinase, gamma-2 subunit; AAKG; AAKG2; adenosine monophosphate-activated protein kinase gamma 2-subunit; AI854673; AMP-activated protein kinase gamma2 subunit; ampk gamma; AMPK gamma 2; AMPK gamma2; AMPK gamma-2 chain; AMPK subunit gamma-2; CMH6; H91620p; OTTHUMP00000211956; PRKAG2; protein kinase AMP-activated non-catalytic subunit gamma 2; protein kinase, AMP-activated, gamma 2 non-catalytic subunit; WPWS |
| Gene Symbols | PRKAG2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.