Learn More
Simport™ Scientific Amplate™ T323 Series Thin Wall PCR Plate
Catalog No. 23045242 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Green
Natural
Yellow
No. of Wells:
24
48
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. of Wells
23-045-242 Green 24
23-045-243 Natural 24
23-045-244 Yellow 24
23-045-245 Natural 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-045-242 Supplier Simport™ Scientific Supplier No. T32324G
Add to Cart
Add to Cart
Available with black alpha-numeric lettering. Amplate™ T323 Series Thin Wall PCR Plate, each well features ultrathin sidewalls for uniform and efficient temperature transfer.
  • Made of polypropylene
  • Available with black alpha-numeric
  • Flexible, can be cut to desired format
  • Autoclavable
  • Inside of tube is smooth and has an inert surface
Certifications
RNase, DNase, Pyrogen and DNA-free.
Specifications
Format Standard
Array 4 x 6
Autoclavable Autoclavable
Color Green
Material Polypropylene
No. of Wells 24
Sterility Non-sterile
No. per Case 50/Cs.