Learn More
Aat Bioquest Amplite™ ADHP [10-Acetyl-3,7-dihydroxyphenoxazine] *CAS#: 119171-73-2*
SDP
Supplier:  Aat Bioquest 11000
Encompass_Preferred
Our Amplite™ ADHP is chemically same to Amplex® Red (Amplex® Red is the trademark of Invitrogen). It is a sensitive fluorogenic peroxidase substrate that has much lower background than the material from other commercial vendors.
Catalog No. 50-195-3325
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.