Learn More
Aat Bioquest Amplite™ IR
SDP
Supplier:  Aat Bioquest 11009
Encompass_Preferred
Our Amplite™ IR is a fluorogenic peroxidase substrate that generates near infrared fluorescence upon reaction with peroxidase and H2O2. It can be used to detect both H2O2 and peroxidase.
Catalog No. 50-195-3331
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.