Learn More
Applied Biosystems™ AmpQuest™ 96-well Real-Time PCR System
Description
- Product Overview:
- Features of the AmpQuest 96 system include:
- 96-well 0.2 mL format
- Factory calibration and no recalibration at fixed time intervals required
- OptiFlex technology (five coupled channels with white LED) for trusted results
- VeriFlex block with six independent programmable temperature zones (25°C range)
- Simplified instrument software with interactive touchscreen for greater ease of use
- Diomni Design and Analysis Software v3 (desktop version)
- Minimized cost of ownership: Fully calibrated and ready for use upon installation, this real-time PCR system supports a wide range of common fluorophores, including FAM, SYBR Green, VIC, ABY, NED, TAMRA, JUN, ROX, Mustang Purple, and Cy5 dyes. Routine recalibration is not required, and flexible warranty extension and service contract options are available to fit different budgets
- Multiplex capabilities minimize consumable/sample requirements: Five coupled excitation and emission filter channels with a white LED enable up to 5-target multiplexing for cost-effective, efficient analysis. OptiFlex technology enhances well-to-well and instrument-to-instrument data accuracy for reliable results
- Performance you can trust: Detect differences in target quantity as small as 1.5-fold, sensitivity down to one copy per reaction, and a linear dynamic range of 10 orders of magnitude
- Feature-rich software designed for research:
- AmpQuest™ 96 Real-Time PCR System is an affordable, high-performance qPCR instrument designed for research and routine testing. Featuring Diomni Design and Analysis Software, a streamlined touchscreen interface and reliable service and support, it delivers accurate data, ease of use and versatility across a wide range of applications
- High application versatility: Simple and optimized protocols, reliable assays and reagents, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, animal health, and more
- System configuration flexibility:
- Option 1:Connect a co-located computer directly to the instrument and send experiments to and from the instrument. This connected computer can be procured along with the AmpQuest 96 system, or you can connect your own laptop or desktop
- Option 2: Network the instrument to allow remote access via a local area network (LAN)
- Option 3: Operate in stand-alone mode (no connections) and transfer files via USB
- Options 2 and 3 do not require a co-located computer
Specifications
Specifications
| Block Format | Non-interchangeable |
| Capacity | 1 x 96-well Plate |
| Voltage | 100/240 V |
| Product Type | Real Time PCR Instrument |
| No. of Programs | 1000 |
| Block Configurations | 96-well |
| Includes | Laptop |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.