Learn More
Applied Biosystems™ AmpQuest™ 96-well Real-Time PCR System
Catalog No. A40012302
Encompass_Preferred
Change view
Click to view available options
Includes:
Desktop
Desktop with Extended Warranty
Extended Warranty
Laptop
Laptop with Extended Warranty
System Only
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
A40012302 Extended Warranty
A40007907 Laptop
A40007906 Desktop
A40007908 System Only
A40012301 Laptop with Extended Warranty
A40012300 Desktop with Extended Warranty
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. A40012302 Supplier Applied Biosystems™ Supplier No. A40012302
Add to Cart
Add to Cart
The system pairs ease-of-use with quality and performance.
  • Product Overview:
    • AmpQuest™ 96 Real-Time PCR System is an affordable, high-performance qPCR instrument designed for research and routine testing. Featuring Diomni Design and Analysis Software, a streamlined touchscreen interface and reliable service and support, it delivers accurate data, ease of use and versatility across a wide range of applications
  • Features of the AmpQuest 96 system include:
    • 96-well 0.2 mL format
    • Factory calibration and no recalibration at fixed time intervals required
    • OptiFlex technology (five coupled channels with white LED) for trusted results
    • VeriFlex block with six independent programmable temperature zones (25°C range)
    • Simplified instrument software with interactive touchscreen for greater ease of use
    • Diomni Design and Analysis Software v3 (desktop version)
  • Minimized cost of ownership:
  • Fully calibrated and ready for use upon installation, this real-time PCR system supports a wide range of common fluorophores, including FAM, SYBR Green, VIC, ABY, NED, TAMRA, JUN, ROX, Mustang Purple, and Cy5 dyes. Routine recalibration is not required, and flexible warranty extension and service contract options are available to fit different budgets
  • Multiplex capabilities minimize consumable/sample requirements:
  • Five coupled excitation and emission filter channels with a white LED enable up to 5-target multiplexing for cost-effective, efficient analysis. OptiFlex technology enhances well-to-well and instrument-to-instrument data accuracy for reliable results
  • Performance you can trust:
  • Detect differences in target quantity as small as 1.5-fold, sensitivity down to one copy per reaction, and a linear dynamic range of 10 orders of magnitude
  • Feature-rich software designed for research:
Diomni Design and Analysis Software v3 (desktop version) is a feature-rich software solution that offers plate setup, quality checks, results review, and a multitude of single-plate application analysis modules
  • High application versatility:
  • Simple and optimized protocols, reliable assays and reagents, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, animal health, and more
  • System configuration flexibility:
    • Option 1:Connect a co-located computer directly to the instrument and send experiments to and from the instrument. This connected computer can be procured along with the AmpQuest 96 system, or you can connect your own laptop or desktop
    • Option 2: Network the instrument to allow remote access via a local area network (LAN)
    • Option 3: Operate in stand-alone mode (no connections) and transfer files via USB
    • Options 2 and 3 do not require a co-located computer
TRUSTED_SUSTAINABILITY
Specifications
Block Format Non-interchangeable
Capacity 1 x 96-well Plate
Voltage 100/240 V
Product Type Real Time PCR Instrument
No. of Programs 1000
Block Configurations 96-well
Includes Extended Warranty