Learn More
Andwin Scientific AMPULE BREAKER/COLLAR 100/PK
Supplier: Andwin Scientific Z122904
The Ampule breaker/collar is compatible with use for 1-4 mL ampules. This product is very easy to use. First, slip the polyethylene breaker/ collar over top of ampule. Then grip the bottom of ampule with one hand and breaker/collar with other. Gently snap top of ampule off. Lastly, discard collar and ampule top. This product is sold in a pack of 100.