Learn More
Abcam AMQI, Fluorogenic Cholinesterase Substrate, Solid, 1 g
Description
- Molecular Weight: 329.13 Da
- Purity: >98%
- Description: Fluorogenic cholinesterase substrate. Hydrolyses to highly fluorescent 7-hydroxy-1-methyl quinolinium iodide.
- Form: Solid
- CAS Number: 7270-83-9
- Composition: 100% (1-methylquinolin-1-ium-7-yl) acetate;iodide
- Storage Temperature: Store at -20°C
- Shipping Temperature: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | Cholinesterase |
| Substrate | AMQI |
| Shipping Condition | Ambient |
| Form | Solid |
| Buffer | 100% (1-methylquinolin-1-ium-7-yl) acetate;iodide |
| Content And Storage | Store at -20°C. |
| Detection Method | Fluorescence |
| Formulation | 100% (1-methylquinolin-1-ium-7-yl) acetate;iodide |
| Quantity | 1 g |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.