Learn More
Abcam AMQI, Fluorogenic Cholinesterase Substrate, Solid, 100 mg
Description
- Properties: Fluorogenic cholinesterase substrate
- Purity: >98%
- Molecular Weight: 329.13 Da
- Form: Solid
- Mechanism: Hydrolyses to highly fluorescent 7-hydroxy-1-methyl quinolinium iodide
- Chemical Constituents: 100% (1-methylquinolin-1-ium-7-yl) acetate;iodide
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Cholinesterase |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Substrate | AMQI |
| Buffer | 100% (1-methylquinolin-1-ium-7-yl) acetate;iodide |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Detection Method | Fluorescence |
| Formulation | 100% (1-methylquinolin-1-ium-7-yl) acetate;iodide |
| Quantity | 100 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.