Learn More
Selleck Chemical LLC Amuvatinib (MP-470) 5mg 850879-09-3 HPK 56
SDP
Supplier:  Selleck Chemical LLC S12445MG
Encompass_Preferred
Amuvatinib (MP-470, HPK 56) is a potent and multi-targeted inhibitor of c-Kit, PDGFR? and Flt3 with IC50 of 10 nM, 40 nM and 81 nM, respectively. Amuvatinib suppresses c-MET and c-RET. Amuvatinib is also active as a DNA repair protein Rad51 inhibitor with antineoplastic activity. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-816-9
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.