Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amygdalin 97.0+%, TCI America™
SDP

Catalog No. a04431g Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A04431G 1 g
A04435G 5 g
A044310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A04431G Supplier TCI America Supplier No. A04431G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29883-15-6
Molecular Formula C20H27NO11
Molecular Weight (g/mol) 457.43
MDL Number MFCD29049822
InChI Key XUCIJNAGGSZNQT-ARGBPYSKNA-N
Synonym amygdalin
PubChem CID 66509032
IUPAC Name 2-phenyl-2-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}acetonitrile
SMILES OC[C@H]1O[C@@H](OC[C@H]2O[C@@H](OC(C#N)C3=CC=CC=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 213°C
Color White
Quantity 1 g
Formula Weight 457.43
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Amygdalin
RTECSNumber OO8450000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.