Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyl Acetate, 98%, Spectrum™ Chemical
SDP

Catalog No. 18608857 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-857 4 L Amber Glass Bottle
18-608-856 25 mL Amber Glass Bottle
18-608-853 100 mL Amber Glass Bottle
18-608-858 500 mL Amber Glass Bottle
18-608-854 20 L Steel Can
18-608-855 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-608-857 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A12804LTGL
Add to Cart
Add to Cart

Chemical Identifiers

CAS 628-63-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
InChI Key PGMYKACGEOXYJE-UHFFFAOYSA-N
IUPAC Name pentyl acetate
SMILES CCCCCOC(C)=O

Specifications

Acidity 0.0001
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 4 L
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.