Learn More
Bachem Amylin (human) Trifluoroacetate
SDP
Supplier:  Bachem 4030200.0500
Encompass_Preferred
Amylin (human) Trifluoroacetate, H-Lys-Cys-Asn-Thr-Ala-Thr-Cys-Ala-Thr-Gln-Arg-Leu-Ala-Asn-Phe-Leu-Val-His-Ser-Ser-Asn-Asn-Phe-Gly-Ala-Ile-Leu-Ser-Ser-Thr-Asn-Val-Gly-Ser-Asn-Thr-Tyr-NH2, 122384-88-7, C165H261N51O55S2, Mr 3903.33, 0.5mg. Islet Amyloid Polypeptide (human), IAPP (human), Amlintide, Amylin, amide, human. The amyloidogenic peptide hormone amylin 1-37 (or Islet Amyloid Polypeptide, IAPP), KCNTATCATQRLANFLVHSSNNFGAILSSTNVGSNTY amide, has been isolated from the amyloid-rich pancreases of diabetic patients. hIAPP forms fibrillar peptide deposits in the pancreatic islets of Langerhans, which may be related to death of the insulin-producing islet b-cells in type 2 diabetes mellitus.
Catalog No. 50-260-168