Learn More
CPC Scientific H-Lys-Cys-Asn-Thr-Ala-Thr-Cys-Ala-Thr-Gln-Arg-Leu-Ala-Asn-Phe-Leu-Val-His-Ser-Ser-Asn-Asn-Phe-Gly-Ala-Ile-Leu-Ser-Ser-Thr-Asn-Val-Gly-Ser-Asn-Thr-Tyr-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific AMYN006B
Encompass
37 amino acid polypeptide secreted from the beta cells of the pancreas and structurally related to calcitonin. It has anorectic effects in rats and may be responsible for the etiology of insulin resistance of type II diabetes mellitus through its modulation of peripheral effects of insulin. It blocks the activation of glycogen synthase by insulin.ONE-LETTER SEQUENCE: KCNTATCATQRLANFLVHSSNNFGAILSSTNVGSNTY-NH2 (C2&C7 bridge)MOLECULAR FORMULA: C165H263N51O55S2MOLECULAR WEIGHT:3903.33STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [122384-88-7], RESEARCH AREA: Diabetes
Catalog No. 50-210-8761
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.