Learn More
Biocare Medical, LLC Amyloid A Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC CM125A
Encompass_Preferred
Amyloidosis is a heterogeneous group of disorders characterized by extracellular deposition of abnormal protein fibrils, which are derived from different proteins. The Amyloid A Antibody reacts with native and fixed amyloid fibrils. The Antibody also reacts with amyloid deposits in many tissues including kidney and rectum. Cross-reactivity with serum precursor of protein AA has been observed. The application of Congo Red, Amyloid A and Amyloid P in tissues with amyloid deposits has been shown to be superior to Congo Red alone.
Catalog No. 50-825-29
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.