Learn More
Bachem Amyloid Beta Protein (1-42) (HFIP-treated)
SDP
Supplier:  Bachem 4090148.0100
Encompass_Preferred
Amyloid Beta Protein (1-42) (HFIP-treated), H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH (HFIP-treated), 107761-42-2, C203H311N55O60S, Mr 4514.10, 0.1mg. Ab42 (HFIP-treated). The peptide was obtained by dissolving Amyloid b-Protein (1-42) in HFIP, aliquoting, and removing the solvent as described in the literature. The morphology of the resulting peptide was studied by transmission electron microscopy (TEM). The magenta arrow points to the nucleation centers where aggregation starts. The yellow arrow indicates the fibrils which are formed as long thin helical structures with regular twists.
Catalog No. NC1079116
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.