Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid beta A4 protein Rat anti-Mouse, Human, Clone: 2E9, MilliporeSigma™

Catalog No. MABN2295MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2295MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2295MI Supplier MilliporeSigma Supplier No. MABN2295
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Amyloid beta A4 protein, clone 2E9 Antibody is a highly specific rat monoclonal antibody that targets Amyloid beta A4 protein and has been tested in Immunofluorescence, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amyloid beta A4 protein
Applications Immunoprecipitation, Western Blot, Immunofluorescence
Classification Monoclonal
Clone 2E9
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols APP,A4,AD1
Host Species Rat
Immunogen KLH-conjugated linear peptide corresponding to 11 amino acids from the C-terminal half of human Amyloid beta A4 protein (APP770).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000475
Test Specificity Clone 2E9 detects multiple isoforms of APP. It targets a sequence of 11 amino acids in the C-terminal region.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.