Learn More
IBL America Amyloid Beta (N) Aβ Anti-Human Rabbit IgG Affinity Purify
Supplier:  IBL America 18584
Encompass_Preferred
Alzheimer's disease (AD) is characterized by the presence of extracellular plaques and intracellular neurofibrillary tangles (NFTs) in the brain. The major protein component of these plaques is beta amyloid (Aβ) peptide, a 40 to 43 amino acid peptide cleaved from amyloid precursor protein by β-secretase and γ-secretase. Increased release of Aβ42 or Aβ43, both of which exhibit a greater tendency to aggregate than Aβ40, occurs in individuals expressing certain genetic mutations, ApoE alleles or may involve other undiscovered factors. Many researchers theorize that it is this increased release of Aβ42/Aβ43 which leads to the abnormal deposition of Aβ and the associated neurotoxicity in the brains of affected individuals. It is also reported that a distinct Aβpeptide, AβN3pE, is deposited in senile plaques in a dominant and differential manner as compared with the standard Aβpeptide. For research use only, not for use in diagnostic procedures.
Catalog No. NC1996392
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.