Learn More
IBL America Amyloid Beta (N3pE-40) Aβ
Supplier:  IBL America 27418
Encompass_Preferred
Solid phase sandwich ELISA for the determination of Human A (N3pE-40) in brain tissue extract. For research use only, not for use in diagnostic procedures.Tetra Methyl Benzidine (TMB) is used as a coloring agent (Chromogen). The strengthof coloring is proportional to the quantities of human A (N3pE-40).
Catalog No. 50-136-6569
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.