Learn More
Apexbio Technology LLC Amyloid Beta-Peptide (1-40) (human), 5mg
SDP
Supplier:  Apexbio Technology LLC A11245
Encompass_Preferred
Amyloid -Peptide (1-40) (human), (C194H295N53O58S1), a peptide with the sequence H2N-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA-OH, MW= 4329.8. Amyloid beta (A or Abeta) is a peptide of 36-43 amino acids that is processed from the Amyloid precursor protein. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4687
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.