Learn More
CPC Scientific H-Asp-Ala-Glu-Phe-Arg-Arg-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific AMYD050A
Encompass
This peptide is the English (H6R) mutation of beta-amyloid, which accelerates fibrillation without increasing protofibril formation. The English and Tottori mutations alter Abeta assembly at its earliest stages, monomer folding and oligomerization, and produce oligomers that are more toxic to cultured neuronal cells than are wild type oligomers. The exchange of His6 by Arg influences the structure of the Cu2+ complex formed by Abeta peptides.ONE-LETTER SEQUENCE: DAEFRRDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVMOLECULAR FORMULA: C194H300N54O58S1MOLECULAR WEIGHT:4348.91STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1802084-26-9], SYNONYMS: [Arg6]-beta-Amyloid (1-40), England MutationRESEARCH AREA: Alzheimer's DiseaseREFERENCES: Y.Hori et al., J. Biol. Chem., 282, 4916 (2007); K.Ono et al., J. Biol. Chem., 285, 23186 (2010); B.Alies et al., Inorg. Chem., 50, 11192 (2011)
Catalog No. 50-210-8710
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.