Learn More
Sigma Aldrich Fine Chemicals Biosciences Amyloid beta Protein Fragm5MG
Supplier: Sigma Aldrich Fine Chemicals Biosciences A1075
Amyloid Protein Fragment 1-40 (A40) is derived from the amyloid- protein (A) which is mapped to human chromosome 21q21.3. A40 is predominantly present in the vascular amyloid deposits. A40 comprises of C-terminal membrane insertion domain. It shows structural transition from random coil to a helical structure in a water-micelle medium.