Learn More
CPC Scientific Glu-Ala-Ser-Asn-Cys-Phe-Ala-Ile-Arg-His-Phe-Glu-Asn-Lys-Phe-Ala-Val-Glu-Thr-Leu-Ile-Cys-Ser (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific AMYD037A
Encompass
ONE-LETTER SEQUENCE: EASNCFAIRHFENKFAVETLICS (Cys5 and 22 bridge)MOLECULAR FORMULA: C116H175N31O35S2MOLECULAR WEIGHT:2628STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [717122-86-6], SYNONYMS: Amyloid Bri Protein Precursor 277 (244-266)RESEARCH AREA: Alzheimer's Disease
Catalog No. 50-210-8695
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.