Learn More
Bachem Amyloid Beta Protein (1-40) Trifluoroacetate
SDP
Supplier:  Bachem 4014442.0500
Encompass_Preferred
Amyloid Beta Protein (1-40) Trifluoroacetate, H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH, 131438-79-4, C194H295N53O58S, Mr 4329.86, 0.5mg. Ab40 trifluoroacetate salt, b-Amyloid (1-40). Amyloid b-peptide (1-40) showed both neurotrophic and neurotoxic effects in dependence on the neuronal age and the concentration of the b-protein. Ab 1-40 has been used as well as Ab 1-42 to detect amyloid b-protein multimers in the cerebrospinal fluid of Alzheimer's disease patients. The sequence of this peptide corresponds to the sequence of human, bovine, canine, feline, ovine, guinea pig, and rabbit Ab40. The inverted sequence can be used as inactive control in experiments employing Ab 1-40. For detailed descriptions of the preparation of wild-type and mutant Ab 1-40 monomers and protofibrils please see the paper of Jan, Hartley, and Lashuel.
Catalog No. 50-194-6568
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.